missing translation for 'onlineSavingsMsg'
Learn More

Ethylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog Number p-7051629
32.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
0.5 mol
1 mol
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
925-90-6
C2H5BrMg
133.27
MFCD00000043
TWTWFMUQSOFTRN-UHFFFAOYSA-M
ethylmagnesium bromide, ethyl magnesium bromide, magnesium, bromoethyl, ethylmagnesiumbromide, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesiumbromid, ethylmagnesium bromid, grignard reagent
101914
magnesium;ethane;bromide
CC[Mg]Br
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
925-90-6
C2H5BrMg
133.27
MFCD00000043
TWTWFMUQSOFTRN-UHFFFAOYSA-M
ethylmagnesium bromide, ethyl magnesium bromide, magnesium, bromoethyl, ethylmagnesiumbromide, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesiumbromid, ethylmagnesium bromid, grignard reagent
101914
magnesium;ethane;bromide
CC[Mg]Br

Ethylmagnesium bromide is used as a reagent and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Melting Point -116.3°C
Color Brown to Yellow
Density 1.020 g/mL
Boiling Point 34.6°C
Odor Ether-like
Packaging ChemSeal™ bottles
Linear Formula C2H5MgBr
Quantity 0.5 mol
UN Number UN3399
Sensitivity Air and moisture sensitive; May form a precipitate
Solubility Information Miscible in alcohols,ketones,esters,ethers,hydrocarbons.
Formula Weight 133.28
Concentration or Composition (by Analyte or Components) 3M in ether
Physical Form Liquid
Chemical Name or Material Ethylmagnesium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'