missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
32.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
0.5 mol
1 mol
Description
Ethylmagnesium bromide is used as a reagent and pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Melting Point | -116.3°C |
Color | Brown to Yellow |
Density | 1.020 g/mL |
Boiling Point | 34.6°C |
Odor | Ether-like |
Packaging | ChemSeal™ bottles |
Linear Formula | C2H5MgBr |
Quantity | 0.5 mol |
UN Number | UN3399 |
Sensitivity | Air and moisture sensitive; May form a precipitate |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'