missing translation for 'onlineSavingsMsg'
Learn More

Ethyl valerate, 99%, Thermo Scientific Chemicals

Catalog Number p-3757922
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
539-82-2
C7H14O2
130.19
MFCD00009479
ICMAFTSLXCXHRK-UHFFFAOYSA-N
ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462
10882
ethyl pentanoate
CCCCC(=O)OCC
This item is not returnable. View return policy
539-82-2
C7H14O2
130.19
MFCD00009479
ICMAFTSLXCXHRK-UHFFFAOYSA-N
ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462
10882
ethyl pentanoate
CCCCC(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8700g/mL
Boiling Point 145°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)3COOC2H5
Refractive Index 1.3990 to 1.401
Quantity 5 g
Merck Index 15, 10090
Specific Gravity 0.87
Formula Weight 130.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl valerate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl valerate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'