missing translation for 'onlineSavingsMsg'
Learn More

Ethyl oxamate, 99%, Thermo Scientific Chemicals

Catalog Number p-3757891
12.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
617-36-7
MFCD00008005
RZMZBHSKPLVQCP-UHFFFAOYSA-N
ethyl oxamate, ethyl carbamoylformate, oxamic acid ethyl ester, ethyl amino oxo acetate, oxamethane, acetic acid, aminooxo-, ethyl ester, ethoxalamide, oxamidic acid ethyl ester, ethoxy carbonyl formamide, oxalic acid, monoethyl ester amide
69238
ethyl 2-amino-2-oxoacetate
CCOC(=O)C(=O)N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
617-36-7
MFCD00008005
RZMZBHSKPLVQCP-UHFFFAOYSA-N
ethyl oxamate, ethyl carbamoylformate, oxamic acid ethyl ester, ethyl amino oxo acetate, oxamethane, acetic acid, aminooxo-, ethyl ester, ethoxalamide, oxamidic acid ethyl ester, ethoxy carbonyl formamide, oxalic acid, monoethyl ester amide
69238
ethyl 2-amino-2-oxoacetate
CCOC(=O)C(=O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 112°C to 115°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Packaging Glass bottle
Molecular Formula C4H7NO3
Linear Formula H2NCOCO2C2H5
Quantity 25 g
Beilstein 02, 544
Solubility Information Solubility in water: soluble
Molecular Weight (g/mol) 117.1
Formula Weight 117.1
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Ethyl oxamate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'