missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl oxalyl chloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Density | 1.2200g/mL |
Boiling Point | 135°C |
Flash Point | 41°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | ClCOCO2C2H5 |
Refractive Index | 1.4160 to 1.418 |
Quantity | 25 mL |
Beilstein | 02, 541 |
Show More |
Safety and Handling
Product Identifier
- Ethyl oxalyl chloride
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'