missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid, tripotassium salt dihydrate, 99+%, ACROS Organics™

Brand:  Acros Organics 301605000

Code : 29

Additional Details : CAS Number : 65501-24-8 Weight : 0.55000kg

Quantity 500g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10552461

  • £224.50 / 500g


Estimated Shipment
18-05-2018
Add to basket

Description

Description

Chelation/Complexation reagent
Specifications

Chemical Identifiers

Ethylenediaminetetraacetic acid, tripotassium salt dihydrate
99+%
(KOCOCH2)2NCH2CH2N(CH2COOK)CH2COOH·2H2O
≥99%
C10H13K3N2O8·2H2O
442.56

Specifications

Ethylenediaminetetraacetic acid, tripotassium salt dihydrate
500g
Authentic
Fine Crystalline Powder
99+%
C10H13K3N2O8·2H2O
442.56
White
65501-24-8
Plastic bottle
≥99%
9% Max. (K.F.)
(KOCOCH2)2NCH2CH2N(CH2COOK)CH2COOH·2H2O
Videos
SDS