missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, tripotassium salt dihydrate, 99+%
55.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
Specifications
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (on dry substance) (Complexometry) |
Packaging | Plastic bottle |
Linear Formula | (KOCOCH2)2NCH2CH2N(CH2COOK)CH2COOH·2H2O |
Quantity | 100 g |
Formula Weight | 442.56 |
Percent Purity | 99+% |
Physical Form | Fine Crystalline Powder |
Chemical Name or Material | Ethylenediaminetetraacetic acid, tripotassium salt dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'