missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
72.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
62.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "22914" to get your promotional price.
62.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
Specifications
Melting Point | 300.0°C |
Assay Percent Range | 99.0 to 102% (on dry substance) (Complexometry) |
Packaging | Plastic bottle |
Quantity | 1 kg |
Solubility Information | (16% in water) Clear solution |
Formula Weight | 416.21 |
Percent Purity | 99% |
Chemical Name or Material | Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'