missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid manganese disodium salt hydrate, Thermo Scientific Chemicals

Catalog Number p-7051128
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
15375-84-5
C10H12MnN2Na2O8
389.13
MFCD00661156
ZQHKAIDDHTYINE-UHFFFAOYSA-J
disodium manganese edta, edta disodium manganese salt, disodium manganese ethylenediaminetetraacetate, manganese disodium ethylene diamine tetraacetate, ethylenediaminetetraacetic acid, disodium manganese salt, disodium ethylenedinitrilo tetraacetato manganese, manganate 2-, disodium, glycine,n'-1,2-ethanediylbis n-carboxymethyl-, manganese disodium salt, manganate 2-,n'-1,2-ethanediylbis n-carboxymethyl glycinato 4--n,n',o,o',on,on'-, disodium, oc-6-21
5148022
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;manganese(2+)
[Na+].[Na+].[Mn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
This item is not returnable. View return policy
15375-84-5
C10H12MnN2Na2O8
389.13
MFCD00661156
ZQHKAIDDHTYINE-UHFFFAOYSA-J
disodium manganese edta, edta disodium manganese salt, disodium manganese ethylenediaminetetraacetate, manganese disodium ethylene diamine tetraacetate, ethylenediaminetetraacetic acid, disodium manganese salt, disodium ethylenedinitrilo tetraacetato manganese, manganate 2-, disodium, glycine,n'-1,2-ethanediylbis n-carboxymethyl-, manganese disodium salt, manganate 2-,n'-1,2-ethanediylbis n-carboxymethyl glycinato 4--n,n',o,o',on,on'-, disodium, oc-6-21
5148022
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;manganese(2+)
[Na+].[Na+].[Mn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Metal chelating compound

Ethylenediaminetetraacetic acid manganese disodium salt hydrate is a metal chelating compound. Cobalt was analysed complexometrically using ethylenediaminetetra-acetic acid (EDTA) by masking aluminum. Previous studies of manganese-EDTA and manganese-EGT-1 complexes indicated that the &-nitrogen coordination of EDTA resulted in a highly anisotropic spectrum for Mn(II). Lipophilic derivatives of manganese EDTA chelates when incorporated into liposomes confer the greatest hepatic enhancement per micromole of metal ion and have favorable clearance kinetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Odor Odorless
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 389.13 (Anhydrous)
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid manganese disodium salt hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'