missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic acid, iron(III) monosodium salt, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
Ethylenediaminetetraacetic acid, iron(III) monosodium salt is used to eliminate enzyme inhibition by traces of heavy metals and to inhibit enzymes that require divalent cations as cofactors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Odor | Odorless |
Quantity | 100 g |
Solubility Information | Soluble in water. |
Formula Weight | 367.05 |
Physical Form | Powder |
Chemical Name or Material | Ethylenediaminetetraacetic acid, iron (III) monosodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'