missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical D/0450/53

Code : 21

Additional Details : CAS Number : 60-00-4 Weight : 0.50000kg


 View more versions of this product

Product Code. 10080060

Quantity Price
1 £82.60 / 500g
4 £74.96 / 500g
16 £71.23 / 500g
In stock, ready to ship: 69

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

60-00-4
3541
Powder Solid
0.013 hPa at 20°C
220°C
500g
C10H16N2O8
292.23g/mol
Plastic powder jar
White
2.5
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethylenediaminetetraacetic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS Documents
Certificates
View accessory products
Special Offers

Special Offers