Learn More
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.1M (0.2N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP , Fisher Chemical™
Brand: Fisher Chemical J/3700/24
Code : 23
Additional Details : CAS Number : 139-33-3 Weight : 10.00000kg
Packaging | Bag in box |
---|---|
Quantity | 10L |
Chemical Identifiers
139-33-3 | |
336.208 | |
ZGTMUACCHSMWAC-UHFFFAOYSA-L | |
57339238 | |
disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate |
C10H14N2Na2O8 | |
70672 | |
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid | |
CHEBI:64734 | |
C(C[NH+](CC(=O)[O-])CC(=O)[O-])[NH+](CC(=O)[O-])CC(=O)[O-].[Na+].[Na+] |
Specifications
Bag in box | |
10L | |
C10H14N2Na2O8 | |
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid | |
C(C[NH+](CC(=O)[O-])CC(=O)[O-])[NH+](CC(=O)[O-])CC(=O)[O-].[Na+].[Na+] | |
336.208 | |
CHEBI:64734 |
Colorless | |
139-33-3 | |
70672 | |
ZGTMUACCHSMWAC-UHFFFAOYSA-L | |
disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate | |
57339238 | |
Liquid |