missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for molecular biology, DNAse, RNAse and Protease free
24.39 GBP valid until 2024-03-30
Use promo code "21648" to get your promotional price.
18.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21648" to get your promotional price.
18.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent for molecular biology
Specifications
Absorbance | (0.1M in water), (0.1 M in water) at 0nm, 0.02 max. at 280nm, 0.2 max. at 260nm |
Melting Point | 252.0°C |
Color | White |
pH | 4 to 5 (5% soln. at 25°C) |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (Complexometry) |
Packaging | Plastic bottle |
Quantity | 100 g |
Merck Index | 153565 |
Solubility Information | Solubility in water: freely soluble. Other solubilities: slightly soluble in alcohol (95%),insoluble in chloroform and ether |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'