missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate, Thermo Scientific Chemicals
15.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is used as chelating agents over a broad pH range in aqueous systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is used as chelating agents over a broad pH range in aqueous systems.
Solubility
Soluble in water. Insoluble in acid and organic liquids
Notes
Incompatible with oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store at room temperature.
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is used as chelating agents over a broad pH range in aqueous systems.
Solubility
Soluble in water. Insoluble in acid and organic liquids
Notes
Incompatible with oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store at room temperature.
Specifications
Melting Point | >300°C |
Odor | Odorless |
Quantity | 25 g |
Solubility Information | Soluble in water. Insoluble in acid and organic liquids |
Formula Weight | 358.50 (Anhydrous) |
Chemical Name or Material | Ethylenediaminetetraacetic acid disodium magnesium salt hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'