missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid, Certified AR for Analysis, Fisher Chemical™
56.70 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
100 g
250 g
Specifications
Melting Point | 220°C |
Color | White |
pH | 2.5 |
Physical Form | Powder/Solid |
Quantity | 100 g |
Formula Weight | 292.23g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 0.013 hPa at 20°C |
Safety and Handling
Product Identifier
- Ethylenediaminetetraacetic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'