missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethylenebis(triphenylphosphonium bromide), 98+%

Brand:  Alfa Aesar™ A12190.14

Code : NEW

Additional Details : CAS Number : 1519-45-5 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11424843

  • £52.05 / 25g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

Ethylenebis (triphenylphosphonium bromide)
C38H34Br2P2
MFCD00031712
1519-45-5
712.45
3901315

Specifications

Ethylenebis (triphenylphosphonium bromide)
≥98%
712.45
3901315
25g
1519-45-5
C38H34Br2P2
MFCD00031712
317° to 320°C
Videos
SDS