missing translation for 'onlineSavingsMsg'
Learn More

Ethylenebis(triphenylphosphine)platinum(0), 98%, Thermo Scientific Chemicals

Catalog Number p-4813480
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
12120-15-9
C38H34P2Pt
747.72
MFCD00012028
RZUASTIKPBCXPU-UHFFFAOYSA-N
ethene bis(triphenylphosphane) platinum
[Pt].C=C.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
12120-15-9
C38H34P2Pt
747.72
MFCD00012028
RZUASTIKPBCXPU-UHFFFAOYSA-N
ethene bis(triphenylphosphane) platinum
[Pt].C=C.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 127.0°C to 131.0°C
Assay Percent Range 97.5% min. (ex Pt)
Packaging Glass bottle
Linear Formula [(C6H5)3P]2Pt[H2C=CH2]
Quantity 250 mg
Solubility Information Solubility in water: insoluble
Formula Weight 747.73
Percent Purity 98%
Chemical Name or Material Ethylenebis(triphenylphosphine)platinum(0)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'