missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene glycol, Reagent Grade, ≥99%, Honeywell™
389.23 GBP valid until 2024-03-31
Use promo code "21663" to get your promotional price.
Use promo code "21663" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
6 x 500 mL
20 L
30 kg
4 x 2.5 L
225 kg
Packaging:
Metal Drum
Poly Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | -13°C (literature) |
CAS Max % | 100.0000% |
Linear Formula | HOCH2CH2OH |
Beilstein | 505945 |
Formula Weight | 62.07 |
Percent Purity | >99% |
Grade | Reagent |
Density | 1.113 g/mL at 25 °C(lit.) |
Boiling Point | 197°C |
Quantity | 20 L |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'