missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog Number p-7059189
Click to view available options
Quantity:
50 g
250 g
1 kg
4 kg
This item is not returnable. View return policy
97-90-5
C10H14O4
198.22
MFCD00008590
STVZJERGLQHEKB-UHFFFAOYSA-N
ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate
7355
CHEBI:53436
2-(2-methylprop-2-enoyloxy)ethyl 2-methylprop-2-enoate
CC(=C)C(=O)OCCOC(=O)C(C)=C
This item is not returnable. View return policy
97-90-5
C10H14O4
198.22
MFCD00008590
STVZJERGLQHEKB-UHFFFAOYSA-N
ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate
7355
CHEBI:53436
2-(2-methylprop-2-enoyloxy)ethyl 2-methylprop-2-enoate
CC(=C)C(=O)OCCOC(=O)C(C)=C

Crosslinking monomer

Ethylene glycol dimethacrylate is used as a functional monomer for polymers and as a cross linking agent between the molecular chains of polymers and elastomers. It is also used in free radical copolymer cross linking reactions. It acts as an intermediate in the production of hydroxyapatite and poly methyl methacrylate composites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.051 g/mL
Boiling Point 83°C to 85°C (1 mmHg)
Flash Point >110°C (230°F)
Linear Formula H2C=C(CH3)CO2CH2CH2OCOC(CH3)=CH2
Refractive Index 1.454
Quantity 50 g
Beilstein 1776663
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 198.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethylene glycol dimethacrylate, Stabilized with 100ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'