missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol diethyl ether, 99%, Thermo Scientific Chemicals

Catalog Number 10581374
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
629-14-1
C6H14O2
118.18
MFCD00009253
LZDKZFUFMNSQCJ-UHFFFAOYSA-N
ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu
12375
1,2-diethoxyethane
CCOCCOCC
This item is not returnable. View return policy
629-14-1
C6H14O2
118.18
MFCD00009253
LZDKZFUFMNSQCJ-UHFFFAOYSA-N
ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu
12375
1,2-diethoxyethane
CCOCCOCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -74.0°C
Color Colorless
Density 0.8417g/mL
Boiling Point 121.0°C
Flash Point 19°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OCH2CH2OC2H5
Refractive Index 1.3910 to 1.3930
Quantity 100 mL
Beilstein 01, 468
Specific Gravity 0.8417
Solubility Information Solubility in water: 20.4%.
Viscosity 0.7 mPa.s (20°C)
Formula Weight 118.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethylene glycol diethyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'