missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethylene glycol diacetate, 97%

Brand:  Alfa Aesar™ 041910.36

Code : NEW

Additional Details : CAS Number : 111-55-7 Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 15441737

  • £19.70 / 500g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

Ethylene glycol diacetate
146.14
1762308
1,2-Diacetoxyethane; Glycol diacetate
111-55-7
MFCD00008718
14,3799

Specifications

Ethylene glycol diacetate
Liquid
146.14
1762308
1,2-Diacetoxyethane; Glycol diacetate
1.13g/mL
−41°C
82° (181°F)
111-55-7
97%
MFCD00008718
14,3799
1.4152
186° to 187°C
500g
Videos
SDS