missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number 15598564
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
107-21-1
C2H6O2
62.068
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
174
CHEBI:30742
ethane-1,2-diol
C(CO)O
This item is not returnable. View return policy
107-21-1
C2H6O2
62.068
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
174
CHEBI:30742
ethane-1,2-diol
C(CO)O

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
Formula Weight 62.07
Grade BAKER ANALYZED™ Reagent
Quantity 500 mL
Flash Point 111.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.10kg
Chemical Name or Material Ethylene Glycol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'