missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene glycol, technical, Thermo Scientific Chemicals
18.97 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -13.0°C |
CAS | 107-21-1 |
Density | 1.1130g/mL |
Boiling Point | 196.0°C to 198.0°C |
Quantity | 1 L |
Flash Point | 116°C |
Refractive Index | 1.4300 to 1.4330 |
Specific Gravity | 1.113 |
Molecular Formula | C2H6O2 |
Linear Formula | HOCH2CH2OH |
Show More |
Safety and Handling
Product Identifier
- Ethylene glycol
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'