missing translation for 'onlineSavingsMsg'
Learn More

Ethylene carbonate, 99+%, Thermo Scientific Chemicals

Catalog Number 10724231
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
96-49-1
C3H4O3
88.06
MFCD00005382
KMTRUDSVKNLOMY-UHFFFAOYSA-N
ethylene carbonate, glycol carbonate, ethylene glycol carbonate, dioxolone-2, cyclic ethylene carbonate, ethylene carbonic acid, 1,3-dioxacyclopentan-2-one, 2-dioxolone, carbonic acid, cyclic ethylene ester, texacar ec
7303
1,3-dioxolan-2-one
C1COC(=O)O1
96-49-1
C3H4O3
88.06
MFCD00005382
KMTRUDSVKNLOMY-UHFFFAOYSA-N
ethylene carbonate, glycol carbonate, ethylene glycol carbonate, dioxolone-2, cyclic ethylene carbonate, ethylene carbonic acid, 1,3-dioxacyclopentan-2-one, 2-dioxolone, carbonic acid, cyclic ethylene ester, texacar ec
7303
1,3-dioxolan-2-one
C1COC(=O)O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY
Melting Point 34.0°C to 37.0°C
Color White to Yellow
Density 1.3210g/mL
Boiling Point 246.7°C
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 250 g
Beilstein 19, 100
Fieser 01,372
Specific Gravity 1.321
Solubility Information Solubility in water: 214g/L (20°C). Other solubilities: soluble in ether,n-butanol and carbon tetra-,chloride,miscible with alcohol,ethyl acetate,benzene and,chloroform
Viscosity 1.92 mPa.s (40°C)
Formula Weight 88.06
Percent Purity 99+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Ethylene carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Ethylene carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'