missing translation for 'onlineSavingsMsg'
Learn More

Ethyl crotonate, 98%, Acros Organics™

Brand:  Acros Organics 409861000

Code : 29

Additional Details : CAS Number : 10544-63-5 Weight : 0.15000kg Transport : UN number : 1862 Chem class : 1 Pack group : II


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10702792

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Ethyl crotonate
98%
CH3CH=CHCO2C2H5
MFCD00009289
98%
C6H10O2
114.14

Specifications

1mg KOH/g max.
100g
0.9180g/mL
Glass bottle
0.918
142.0° to 143.0°C
Liquid
98%
CH3CH=CHCO2C2H5
MFCD00009289
Ethyl crotonate
10544-63-5
Authentic
1.4232 to 1.4272
Clear colorless
2°C
98%
C6H10O2
114.14
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethyl crotonate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS