missing translation for 'onlineSavingsMsg'
Learn More

Ethyl crotonate, 98%, Acros Organics™

Brand:  Acros Organics 409860050

Code : 29

Additional Details : CAS Number : 10544-63-5 Weight : 0.05500kg Transport : UN number : 1862 Chem class : 3 Pack group : II


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10415492

  • £8.40 / 5g


Estimated Shipment
11-07-2018
Add to basket


Specifications

Chemical Identifiers

Ethyl crotonate
98%
CH3CH=CHCO2C2H5
MFCD00009289
98%
C6H10O2
114.14

Specifications

1mg KOH/g max.
5g
0.9180g/mL
Glass bottle
0.918
142.0° to 143.0°C
Liquid
98%
CH3CH=CHCO2C2H5
MFCD00009289
Ethyl crotonate
10544-63-5
Authentic
1.4232 to 1.4272
Clear colorless
2°C
98%
C6H10O2
114.14
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethyl crotonate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS