missing translation for 'onlineSavingsMsg'
Learn More

Ethyl caproate, 99+%, Thermo Scientific Chemicals

Catalog Number 10561801
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
123-66-0
C8H16O2
144.21
MFCD00009511
SHZIWNPUGXLXDT-UHFFFAOYSA-N
ethyl caproate, ethyl hexoate, hexanoic acid, ethyl ester, hexanoic acid ethyl ester, capronic ether absolute, caproic acid ethyl ester, ethyl n-hexanoate, ethyl butyl acetate, caproate, acetic acid, butyl-, ethyl ester
31265
CHEBI:86055
ethyl hexanoate
CCCCCC(=O)OCC
This item is not returnable. View return policy
123-66-0
C8H16O2
144.21
MFCD00009511
SHZIWNPUGXLXDT-UHFFFAOYSA-N
ethyl caproate, ethyl hexoate, hexanoic acid, ethyl ester, hexanoic acid ethyl ester, capronic ether absolute, caproic acid ethyl ester, ethyl n-hexanoate, ethyl butyl acetate, caproate, acetic acid, butyl-, ethyl ester
31265
CHEBI:86055
ethyl hexanoate
CCCCCC(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 1mg KOH/g max.
Melting Point 67°C
Color Colorless
Density 0.8690g/mL
Boiling Point 168°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CO2C2H5
Refractive Index 1.4060 to 1.409
Quantity 500 mL
Beilstein 02, 323
Merck Index 15, 3832
Specific Gravity 0.869
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in glycerol,soluble in alcohol and ether
Formula Weight 144.21
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl caproate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl caproate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'