missing translation for 'onlineSavingsMsg'
Learn More

Ethyl bromopyruvate, 80-85%, Thermo Scientific Chemicals

Catalog Number 10500751
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
70-23-5
C5H7BrO3
195.01
MFCD00000204
VICYTAYPKBLQFB-UHFFFAOYSA-N
ethyl bromopyruvate, ethyl 3-bromopyruvate, propanoic acid, 3-bromo-2-oxo-, ethyl ester, bromopyruvic acid ethyl ester, ethyl bromo pyruvate, ethyl 3-bromo-2-oxopropionate, pyruvic acid, bromo-, ethyl ester, .beta.-bromopyruvic acid ethyl ester, 3-bromo-2-oxo-propionic acid ethyl ester, beta-bromopyruvic acid ethyl ester
66144
ethyl 3-bromo-2-oxopropanoate
CCOC(=O)C(=O)CBr
This item is not returnable. View return policy
70-23-5
C5H7BrO3
195.01
MFCD00000204
VICYTAYPKBLQFB-UHFFFAOYSA-N
ethyl bromopyruvate, ethyl 3-bromopyruvate, propanoic acid, 3-bromo-2-oxo-, ethyl ester, bromopyruvic acid ethyl ester, ethyl bromo pyruvate, ethyl 3-bromo-2-oxopropionate, pyruvic acid, bromo-, ethyl ester, .beta.-bromopyruvic acid ethyl ester, 3-bromo-2-oxo-propionic acid ethyl ester, beta-bromopyruvic acid ethyl ester
66144
ethyl 3-bromo-2-oxopropanoate
CCOC(=O)C(=O)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Pink-Red to Yellow
Density 1.5500g/mL
Boiling Point 100.0°C to 102.0°C (15.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2COCO2C2H5
Refractive Index 1.4675 to 1.4765
Quantity 5 mL
Beilstein 03, II, 409
Specific Gravity 1.55
Formula Weight 195.01
Percent Purity 80-85%
Physical Form Liquid
Chemical Name or Material Ethyl bromopyruvate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl bromopyruvate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'