missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle6-chloroimidazo[1,2-b]pyridazine-2-carboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 225.63 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 6-chloroimidazo[1, 2-b]pyridazine-2-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'