missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-(chloromethyl)-2-furancarboxylate, 95%, Acros Organics™

Brand:  Acros Organics 349730050

Code : 29

Additional Details : CAS Number : 2528-00-9 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10461972

  • £59.50 / 5g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

Ethyl 5-(chloromethyl)-2-furancarboxylate
95%
C8H9ClO3
MFCD00173837
2528-00-9
95%
188.61

Specifications

Ethyl 5-(chloromethyl)-2-furancarboxylate
1.2250g/mL
Glass bottle
1.225
95%
115.0°C
C8H9ClO3
>110°C
MFCD00173837
2528-00-9
Authentic
1.5100 to 1.5130
Liquid
95%
Clear yellow
5g
188.61
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethyl 5-(chloromethyl)-2-furancarboxylate

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.

Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.

SDS