missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-aminobenzo[b]furan-2-carboxylate, 97%, Thermo Scientific™

Catalog Number p-4228647
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
174775-48-5
C11H11NO3
205.21
MFCD08275091
YFFLLDHEEWSHQG-UHFFFAOYSA-N
ethyl 5-aminobenzofuran-2-carboxylate, 2-benzofurancarboxylic acid, 5-amino-, ethyl ester, ethyl 5-aminobenzo b furan-2-carboxylate, 5-amino-2-ethoxycarbonyl-1-benzofuran, 5-aminobenzofuran-2-carboxylic acid ethyl ester, 5-amino-benzofuran-2-carboxylic acid ethyl ester, ethyl 5-aminobenzo d furan-2-carboxylate, pubchem23808, ethylaminobenzofurancarboxylate, ksc494o5f
9837138
ethyl 5-amino-1-benzofuran-2-carboxylate
CCOC(=O)C1=CC2=CC(N)=CC=C2O1
This item is not returnable. View return policy
174775-48-5
C11H11NO3
205.21
MFCD08275091
YFFLLDHEEWSHQG-UHFFFAOYSA-N
ethyl 5-aminobenzofuran-2-carboxylate, 2-benzofurancarboxylic acid, 5-amino-, ethyl ester, ethyl 5-aminobenzo b furan-2-carboxylate, 5-amino-2-ethoxycarbonyl-1-benzofuran, 5-aminobenzofuran-2-carboxylic acid ethyl ester, 5-amino-benzofuran-2-carboxylic acid ethyl ester, ethyl 5-aminobenzo d furan-2-carboxylate, pubchem23808, ethylaminobenzofurancarboxylate, ksc494o5f
9837138
ethyl 5-amino-1-benzofuran-2-carboxylate
CCOC(=O)C1=CC2=CC(N)=CC=C2O1
Melting Point 56°C to 64°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 205.22
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Ethyl 5-aminobenzo[b]furan-2-carboxylate
compliance-icons
Product Identifier
  • Ethyl 5-aminobenzo¢b!furan-2-carboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'