missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle4-nitrocinnamate, 99%, Predominantly trans, ACROS Organics™
Click to view available options
25g; Glass bottle
Specifications
CAS | 953-26-4 |
Melting Point | 137°C to 140°C |
Color | Beige-Yellow to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Molecular Formula | C11H11NO4 |
Linear Formula | O2NC6H4CH=CHCO2C2H5 |
MDL Number | MFCD00007380 |
Quantity | 25 g |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'