missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-methyl-4-pentenoate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.8910g/mL |
Boiling Point | 85.0°C (20.0 mmHg) |
Flash Point | 47°C |
Assay Percent Range | 94% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH2=C(CH3)CH2CH2C(O)OCH2CH3 |
Quantity | 5 g |
Specific Gravity | 0.891 |
Formula Weight | 142.2 |
Percent Purity | 95% |
Show More |
Safety and Handling
Product Identifier
- Ethyl 4-methyl-4-pentenoate
- Warning
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'