missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
64.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 115°C to 118°C |
Color | White |
Boiling Point | 297°C to 298°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Plastic bottle |
Linear Formula | HOC6H4CO2C2H5 |
Quantity | 1 kg |
Beilstein | 10, 159 |
Merck Index | 15, 3890 |
Show More |
Safety and Handling
Product Identifier
- Ethyl 4-hydroxybenzoate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'