missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle4-formyl-3,5-dimethyl-1H-pyrrole-2-carboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 195.22 |
Percent Purity | 97% |
Chemical Name or Material | ethyle4-formyl-3, 5-dimethyl-1H-pyrrole-2-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'