missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 50 g |
Formula Weight | 246.1 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 4-bromo-3, 5-dimethyl-1H-pyrrole-2-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'