missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-bromobenzoate, 98%, Thermo Scientific Chemicals

Catalog Number p-3778551
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
24398-88-7
C9H9BrO2
229.08
MFCD00013529
QAUASTLEZAPQTB-UHFFFAOYSA-N
90488
ethyl 3-bromobenzoate
CCOC(=O)C1=CC(=CC=C1)Br
This item is not returnable. View return policy
24398-88-7
C9H9BrO2
229.08
MFCD00013529
QAUASTLEZAPQTB-UHFFFAOYSA-N
90488
ethyl 3-bromobenzoate
CCOC(=O)C1=CC(=CC=C1)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.4300g/mL
Boiling Point 130°C to 131°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CO2C2H5
Refractive Index 1.5405 to 1.5425
Quantity 5 g
Specific Gravity 1.43
Synonym ethyl3-bromobenzoate, 3-bromobenzoic acid ethyl ester, benzoic acid, 3-bromo-, ethyl ester, ethyl-3-bromobenzoate, benzoic acid, m-bromo-, ethyl ester, m-bromo benzoic acid ethyl ester, m-bromobenzoic acid, ethyl ester, pubchem3941, acmc-209gbv, ethyl 3-bromobenzoate
Formula Weight 229.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 3-bromobenzoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'