missing translation for 'onlineSavingsMsg'
Learn More

ethyle3-bromobenzoate, 98%, ACROS Organics™

Brand:  Acros Organics 308540250

Code : 29

Additional Details : CAS Number : 24398-88-7 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10572471

  • £52.80 / 25g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

ethyle3-bromobenzoate
98%
BrC6H4COCH5
MFCD00013529
≥97.5%
C9H9BrO2
229.08

Specifications

ethyle3-bromobenzoate
1.43g/mL
Glass bottle
Liquid
98%
C9H9BrO2
BrC6H4COCH5
MFCD00013529
24398-88-7
Authentic
1.5405 to 1.5425 (20°C, 589nm)
≥97.5%
Clear colorless
25g
229.08
Videos
SDS