missing translation for 'onlineSavingsMsg'
Learn More

ethyle3-aminobenzoate, +99%, ACROS Organics™

Brand:  Acros Organics 117991000

Code : 29

Additional Details : CAS Number : 582-33-2 Weight : 0.15000kg


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10561564

  • £192.60 / 100g


Estimated Shipment
18-06-2018
Add to basket


Specifications

Chemical Identifiers

Ethyl 3-aminobenzoate
99+%
H2NC6H4CO2C2H5
MFCD00007794
≥99.0%
C9H11NO2
165.19

Specifications

Ethyl 3-aminobenzoate
Clear yellow to light brown
582-33-2
Glass bottle
Liquid or Low Melting Solid
99+%
H2NC6H4CO2C2H5
MFCD00007794
1.1070g/mL
100g
Authentic
1.5595 to 1.5615 (20°C, 589nm)
≥99.0%
C9H11NO2
165.19
14, 389
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethyl 3-aminobenzoate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS