missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methylnicotinate, 97%, Thermo Scientific™

Catalog Number 10421250
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1721-26-2
C9H11NO2
165.19
MFCD00006336
TZORNSWZUZDUCU-UHFFFAOYSA-N
ethyl 2-methylnicotinate, 2-methylnicotinic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, ethyl 2-methylpyridine-3-carboxylate∼2-methylnicotinic acid ethyl ester, pubchem12727
74401
ethyl 2-methylpyridine-3-carboxylate
CCOC(=O)C1=CC=CN=C1C
This item is not returnable. View return policy
1721-26-2
C9H11NO2
165.19
MFCD00006336
TZORNSWZUZDUCU-UHFFFAOYSA-N
ethyl 2-methylnicotinate, 2-methylnicotinic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, ethyl 2-methylpyridine-3-carboxylate∼2-methylnicotinic acid ethyl ester, pubchem12727
74401
ethyl 2-methylpyridine-3-carboxylate
CCOC(=O)C1=CC=CN=C1C
Color Colorless to Orange
Density 1.0700g/mL
Boiling Point 126°C to 127°C (24.0 mmHg)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.07
Formula Weight 165.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 2-methylnicotinate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 2-methylnicotinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'