Learn More
Thermo Scientific Maybridge Ethyl 2-methyl-1,3-thiazole-4-carboxylate, 25g, 97%
Brand: Thermo Scientific Maybridge SPB02288EB
Ethyl 2-methyl-1,3-thiazole-4-carboxylate, 25g, 97%
Code : 31
Additional Details : CAS Number : 6436-59-5 Weight : 0.07500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III