missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-chloro-3-cyano-6-methylisonicotinate, 97%, Thermo Scientific™

Catalog Number p-2704108
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
40108-12-1
C10H9ClN2O2
224.644
MFCD00023416
XUXBHWPRQGVLRY-UHFFFAOYSA-N
ethyl 2-chloro-3-cyano-6-methylisonicotinate, ethyl-2-chloro-3-cyano-6-methylisonicotinate, isonicotinic acid, 2-chloro-3-cyano-6-methyl-, ethyl ester, 2-chloro-3-cyano-6-methylisonicotinic acid ethyl ester, 2-chloro-3-cyano-4-carbethoxy-6-methylpyridine, ethyl2-chloro-3-cyano-6-methylisonicotinate, 2-chloro-3-cyano-6-methyl-isonicotinic acid ethyl ester, 4-pyridinecarboxylic acid, 2-chloro-3-cyano-6-methyl-, ethyl ester, 4-pyridinecarboxylicacid, 2-chloro-3-cyano-6-methyl-, ethyl ester
142408
ethyl 2-chloro-3-cyano-6-methylpyridine-4-carboxylate
CCOC(=O)C1=CC(=NC(=C1C#N)Cl)C
This item is not returnable. View return policy
40108-12-1
C10H9ClN2O2
224.644
MFCD00023416
XUXBHWPRQGVLRY-UHFFFAOYSA-N
ethyl 2-chloro-3-cyano-6-methylisonicotinate, ethyl-2-chloro-3-cyano-6-methylisonicotinate, isonicotinic acid, 2-chloro-3-cyano-6-methyl-, ethyl ester, 2-chloro-3-cyano-6-methylisonicotinic acid ethyl ester, 2-chloro-3-cyano-4-carbethoxy-6-methylpyridine, ethyl2-chloro-3-cyano-6-methylisonicotinate, 2-chloro-3-cyano-6-methyl-isonicotinic acid ethyl ester, 4-pyridinecarboxylic acid, 2-chloro-3-cyano-6-methyl-, ethyl ester, 4-pyridinecarboxylicacid, 2-chloro-3-cyano-6-methyl-, ethyl ester
142408
ethyl 2-chloro-3-cyano-6-methylpyridine-4-carboxylate
CCOC(=O)C1=CC(=NC(=C1C#N)Cl)C
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 224.65
Percent Purity 97%
Chemical Name or Material ethyle2-chloro-3-cyano-6-methylisonicotinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'