missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle2-bromobenzoate, 97%, Maybridge™
Click to view available options
Quantity:
10 g
50 g
100 g
Specifications
CAS Min % | 97.0 |
Density | 1.44g/mL |
Assay Percent Range | 97% |
Linear Formula | BrC6H4CO2C2H5 |
Quantity | 100 g |
Formula Weight | 229.07 |
Percent Purity | 97% |
Chemical Name or Material | ethyle2-bromobenzoate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'