missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-thiazoleacetate, 99%, Acros Organics™

Brand:  Acros Organics 205300500

Code : 29

Additional Details : CAS Number : 53266-94-7 Weight : 0.10000kg

Quantity 50g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10772821

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Ethyl 2-amino-4-thiazoleacetate
99%
C7H10N2O2S
MFCD00005330
53266-94-7
99%
186.23

Specifications

Ethyl 2-amino-4-thiazoleacetate
Authentic
Crystalline Powder
91.0° to 95.0°C
Slightly yellow to beige
50g
186.23
53266-94-7
Plastic bottle
99%
99%
C7H10N2O2S
185°C
MFCD00005330
Videos
SDS