Learn More
Thermo Scientific Maybridge Ethyl 2-(4-chlorophenoxy)-2-methylpropanoate, 50g, 97%
Brand: Thermo Scientific Maybridge RDP00167EE
Ethyl 2-(4-chlorophenoxy)-2-methylpropanoate, 50g, 97%
Code : 31
Additional Details : CAS Number : 637-07-0 Weight : 0.10000kg