missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-(3-oxo-2-piperazinyl)acetate, 97%, Thermo Scientific™

Catalog Number 10622063
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
33422-35-4
C8H14N2O3
186.211
MFCD00052897
HNYRNJAZRKCHSC-UHFFFAOYSA-N
ethyl 2-3-oxopiperazin-2-yl acetate, ethyl 2-3-oxo-2-piperazinyl acetate, 2-piperazineacetic acid, 3-oxo-, ethyl ester, 3-oxo-piperazin-2-yl-acetic acid ethyl ester, ethyl 3-oxopiperazine-2-acetate, ethyl 3-oxo-2-piperazinyl acetate, ethyl 3-oxopiperazin-2-yl acetate, ethyl 2s-3-oxopiperazin-2-yl acetate, pubchem17782, ethyloxopiperazinylacetate
544550
ethyl 2-(3-oxopiperazin-2-yl)acetate
CCOC(=O)CC1C(=O)NCCN1
This item is not returnable. View return policy
33422-35-4
C8H14N2O3
186.211
MFCD00052897
HNYRNJAZRKCHSC-UHFFFAOYSA-N
ethyl 2-3-oxopiperazin-2-yl acetate, ethyl 2-3-oxo-2-piperazinyl acetate, 2-piperazineacetic acid, 3-oxo-, ethyl ester, 3-oxo-piperazin-2-yl-acetic acid ethyl ester, ethyl 3-oxopiperazine-2-acetate, ethyl 3-oxo-2-piperazinyl acetate, ethyl 3-oxopiperazin-2-yl acetate, ethyl 2s-3-oxopiperazin-2-yl acetate, pubchem17782, ethyloxopiperazinylacetate
544550
ethyl 2-(3-oxopiperazin-2-yl)acetate
CCOC(=O)CC1C(=O)NCCN1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material ethyle2-(3-oxo-2-piperazinyl)acetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'