missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle2-(2-methyl-1,3-thiazol-4-yl)acetate, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 5 g |
Formula Weight | 185.25 |
Percent Purity | 95% |
Chemical Name or Material | Ethyl 2-(2-methyl-1, 3-thiazol-4-yl)acetate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'