missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-aminocyclopropanecarboxylate hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-4228646
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
42303-42-4
C6H11NO2·ClH
165.62
MFCD00190747
XFNUTZWASODOQK-UHFFFAOYSA-N
ethyl 1-aminocyclopropanecarboxylate hydrochloride, 1-aminocyclopropane-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet.hcl, ethyl 1-aminocyclopropane-1-carboxylate hydrochloride, 1-amino-cyclopropyl-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet hcl, 1-aminocyclopropane-1-carboxylic acid ethyl ester, acpc-oet-hcl, h-acpc-oet?cl, pubchem14733
386203
ethyl 1-aminocyclopropane-1-carboxylate;hydrochloride
CCOC(=O)C1(CC1)N.Cl
This item is not returnable. View return policy
42303-42-4
C6H11NO2·ClH
165.62
MFCD00190747
XFNUTZWASODOQK-UHFFFAOYSA-N
ethyl 1-aminocyclopropanecarboxylate hydrochloride, 1-aminocyclopropane-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet.hcl, ethyl 1-aminocyclopropane-1-carboxylate hydrochloride, 1-amino-cyclopropyl-1-carboxylic acid ethyl ester hydrochloride, h-acpc-oet hcl, 1-aminocyclopropane-1-carboxylic acid ethyl ester, acpc-oet-hcl, h-acpc-oet?cl, pubchem14733
386203
ethyl 1-aminocyclopropane-1-carboxylate;hydrochloride
CCOC(=O)C1(CC1)N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 113.0°C to 114.0°C
Color White
Elemental Analysis 42.41 to 45.48% C, 8.24 to 8.66% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 165.62
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 1-aminocyclopropanecarboxylate hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 1-aminocyclopropanecarboxylate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'