Learn More
Thermo Scientific Maybridge Ethyl 1,4-dibenzylpiperazine-2-carboxylate, 10g, 97%
Brand: Thermo Scientific Maybridge KM07907EA
Ethyl 1,4-dibenzylpiperazine-2-carboxylate, 10g, 97%
Code : 31
Additional Details : CAS Number : 72351-59-8 Weight : 0.06000kg