missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle1-(3-aminobenzyl)piperidine-4-carboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Synonym | ethyl 1-3-aminobenzyl piperidine-4-carboxylate, ethyl 1-3-aminophenyl methyl piperidine-4-carboxylate, 4-piperidinecarboxylicacid, 1-3-aminophenyl methyl-, ethyl ester, ethyl1-3-aminobenzyl piperidine-4-carboxylate, 1-3-aminobenzyl-4-ethoxycarbonyl piperidine, 3-4-ethoxycarbonyl piperidin-1-yl methyl aniline, 1-3-aminophenyl methyl-4-piperidinecarboxylic acid ethyl ester |
Formula Weight | 262.35 |
Percent Purity | 97% |
Chemical Name or Material | ethyle1-(3-aminobenzyl)piperidine-4-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'